6-Bromoveratraldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromo-3,4-dimethoxybenzaldehyde, 2-Bromo-4,5-dimethoxybenzaldehyde
OR27568-100G OR27568-25G
APOSOR27568-100GEA 109 EUR
APOSOR27568-100G APOSOR27568-25G
6-Bromoveratraldehyde
6-Bromoveratraldehyde
Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00003301
CAS Number: 5392-10-9