6-Amino-5-bromonicotinonitrile
Supplier: Apollo Scientific
Warning
OR2759-1G OR2759-5G
APOSOR2759-1GEA 94.4 EUR
APOSOR2759-1G APOSOR2759-5G
6-Amino-5-bromonicotinonitrile
6-Amino-5-bromonicotinonitrile
Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
Melting Pt: 194…195 °C
Storage Temperature: Ambient
MDL Number: MFCD02102412
CAS Number: 477871-32-2