2,4,5-Trimethylaniline
Supplier: Apollo Scientific
Danger
OR27602-1G OR27602-250MG OR27602-5G
APOSOR27602-1GEA 72.5 EUR
APOSOR27602-1G APOSOR27602-250MG APOSOR27602-5G
2,4,5-Trimethylaniline
2,4,5-Trimethylaniline
Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 234
Melting Pt: 68 °C
Density: 0.957
Storage Temperature: Refrigerator
MDL Number: MFCD00025201
CAS Number: 137-17-7
EINECS: 205-282-0
UN: 2811
ADR: 6.1 III