Ethyl-3-oxocyclohexanecarboxylate 97+%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 3-oxocyclohexane-1-carboxylate
OR27815-1G OR27815-250MG
APOSOR27815-1GEA 0 EUR
APOSOR27815-1G APOSOR27815-250MG
Ethyl-3-oxocyclohexanecarboxylate 97+%
Ethyl 3-oxocyclohexanecarboxylate
Formula: C₉H₁₄O₃
Boiling Pt: 135…137 °C/15mmHg
Density: 1.068 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00205583
CAS Number: 33668-25-6