3-Amino-5-(2-furyl)-1H-pyrazole
Supplier: Apollo Scientific
Warning
Synonyms: 5-(Fur-2-yl)-1H-pyrazol-3-amine, 5-(Furan-2-yl)-1H-pyrazol-3-amine
OR27866-1G OR27866-250MG
APOSOR27866-1GEA 87 EUR
APOSOR27866-1G APOSOR27866-250MG
3-Amino-5-(2-furyl)-1H-pyrazole
3-Amino-5-(2-furyl)-1H-pyrazole
Formula: C₇H₇N₃O
MW: 149.15 g/mol
Melting Pt: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00082696
CAS Number: 96799-02-9