2-Amino-1-methylbenzimidazole
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-1H-benzo[d]imidazol-2-amine
OR27921-1G OR27921-5G OR27921-10G
APOSOR27921-1GEA 21.9 EUR
APOSOR27921-1G APOSOR27921-5G APOSOR27921-10G
2-Amino-1-methylbenzimidazole
2-Amino-1-methylbenzimidazole
Formula: C₈H₉N₃
MW: 147.181 g/mol
Melting Pt: 203…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00142855
CAS Number: 1622-57-7