3-(Chloromethyl)benzoyl chloride 93%
Supplier: Apollo Scientific
Danger
OR2797-25G
APOSOR2797-25GEA 171 EUR
APOSOR2797-25G
3-(Chloromethyl)benzoyl chloride 93%
3-(Chloromethyl)benzoyl chloride
Formula: C₈H₆Cl₂O
MW: 189.04 g/mol
Boiling Pt: 149…150 °C/20mmHg
Melting Pt: 4 °C
Density: 1.33 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000682
CAS Number: 63024-77-1
EINECS: 263-799-7
UN: 3265
ADR: 8,II