2-Bromo-5-nitrobenzoic acid 98%
Supplier: Apollo Scientific
OR27973-100G OR27973-25G
APOSOR27973-100GEA 223 EUR
APOSOR27973-100G APOSOR27973-25G
2-Bromo-5-nitrobenzoic acid 98%
2-Bromo-5-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MW: 246.02 g/mol
Melting Pt: 180…181 °C
Storage Temperature: Ambient
MDL Number: MFCD00134558
CAS Number: 943-14-6