Bis(2-chloroethyl)ammonium chloride 98%
Supplier: Apollo Scientific
Danger
Synonyms: Bis(2-chloroethyl)amine hydrochloride
OR27981-10KG OR27981-1KG OR27981-250G
APOSOR27981-10KGEA 545 EUR
APOSOR27981-10KG APOSOR27981-1KG APOSOR27981-250G
Bis(2-chloroethyl)ammonium chloride 98%
Bis(2-chloroethyl)ammonium chloride
Formula: C₄H₁₀Cl₃N
MW: 178.49 g/mol
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012515
CAS Number: 821-48-7
EINECS: 212-479-5
UN: 2923
ADR: 8+6.1 II