3-Chloro-2-nitropyridine 98%
Supplier: Apollo Scientific
OR2799-1G OR2799-5G
APOSOR2799-1GEA 114 EUR
APOSOR2799-1G APOSOR2799-5G
3-Chloro-2-nitropyridine 98%
3-Chloro-2-nitropyridine
Formula: C₅H₃ClN₂O₂
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00490110
CAS Number: 54231-32-2