6-Methyl-1-indanone
Supplier: Apollo Scientific
Warning
OR28016-10G OR28016-50G
APOSOR28016-10GEA 31.9 EUR
APOSOR28016-10G APOSOR28016-50G
6-Methyl-1-indanone
6-Methyl-1-indanone
Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Boiling Pt: 70 °C/0.4mmHg
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00275729
CAS Number: 24623-20-9