Isoxazole-5-carbonyl chloride 97%
Supplier: Apollo Scientific
Danger
OR2811-1G OR2811-250MG OR2811-5G
APOSOR2811-1GEA 46.4 EUR
APOSOR2811-1G APOSOR2811-250MG APOSOR2811-5G
Isoxazole-5-carbonyl chloride 97%
Isoxazole-5-carbonyl chloride
Formula: C₄H₂ClNO₂
MW: 131.52 g/mol
Boiling Pt: 167…168 °C
Density: 1.362 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00067868
CAS Number: 62348-13-4
UN: 3265
ADR: 8 II