Isoquinoline-4-amine
Supplier: Apollo Scientific
Warning
Synonyms: 4-Aminoisoquinoline, Isoquinolin-4-amine
OR2818-1G OR2818-5G
APOSOR2818-1GEA 36.4 EUR
APOSOR2818-1G APOSOR2818-5G
Isoquinoline-4-amine
Isoquinoline-4-amine
Formula: C₉H₈N₂
MW: 144.1700 g/mol
Melting Pt: 107…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00034752
CAS Number: 23687-25-4
EINECS: 245-823-8