2'-Hydroxy-6'-methoxyacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-Hydroxy-6-methoxyphenyl)ethanone
OR28267-10G OR28267-25G OR28267-5G
APOSOR28267-10GEA 74.1 EUR
APOSOR28267-10G APOSOR28267-25G APOSOR28267-5G
2'-Hydroxy-6'-methoxyacetophenone 98%
2'-Hydroxy-6'-methoxyacetophenone
Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 141 °C/16mmHg
Melting Pt: 56…60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008732
CAS Number: 703-23-1
EINECS: 211-872-9