2,4-Dimethoxybenzyl alcohol
Supplier: Apollo Scientific
Warning
OR28314-100G OR28314-25G OR28314-500G
APOSOR28314-100GEA 119 EUR
APOSOR28314-100G APOSOR28314-25G APOSOR28314-500G
2,4-Dimethoxybenzyl alcohol
2,4-Dimethoxybenzyl alcohol
Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Boiling Pt: 177…179 °C/10mmHg
Melting Pt: 38…40 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004614
CAS Number: 7314-44-5
EINECS: 230-775-2