3-Chloro-4-iodoaniline
Supplier: Apollo Scientific
|
Danger
|
OR28394-100G
OR28394-25G
APOSOR28394-100GEA
490
EUR
APOSOR28394-100G
APOSOR28394-25G
3-Chloro-4-iodoaniline
3-Chloro-4-iodoaniline
Formula:
C₆H₅ClIN MW: 253.47 g/mol Melting Pt: 56…58 °C Density: 2.015 g/cm³ (25 °C) Flash Pt: 143 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00051845 CAS Number: 135050-44-1 UN: 2811 ADR: 6.1,III |