Thiodiglycolic anhydride
Supplier: Apollo Scientific
Warning
Synonyms: 1,4-Oxathiane-2,6-dione
OR28488-100G OR28488-25G OR28488-5G
APOSOR28488-100GEA 161 EUR
APOSOR28488-100G APOSOR28488-25G APOSOR28488-5G
Thiodiglycolic anhydride
Thiodiglycolic anhydride
Formula: C₄H₄O₃S
MW: 132.13 g/mol
Melting Pt: 94…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00051689
CAS Number: 3261-87-8