2,6-Dichlorobenzoyl chloride 98%
Supplier: Apollo Scientific
Synonyms: 2,6-Dichlorobenzoyl Chloride
OR2852-100G
APOSOR2852-100GEA 91.5 EUR
APOSOR2852-100G
2,6-Dichlorobenzoyl chloride 98%
2,6-Dichlorobenzoyl chloride
Formula: C₇H₃Cl₃O
MW: 209.46 g/mol
Boiling Pt: 142…143 °C/21mmHg
Melting Pt: 15…18 °C
Density: 1.463 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000662
CAS Number: 4659-45-4
UN: 3265
ADR: 8,II