3,4-(Methylenedioxy)aniline, Technical Grade
Supplier: Apollo Scientific
Warning
Synonyms: 5-Amino-1,3-benzodioxole
OR28575-25G OR28575-100G OR28575-5G
APOSOR28575-25GEA 36.4 EUR
APOSOR28575-25G APOSOR28575-100G APOSOR28575-5G
3,4-(Methylenedioxy)aniline, Technical Grade
3,4-(Methylenedioxy)aniline
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Boiling Pt: 144 °C/16mmHg
Melting Pt: 40…45 °C
Flash Pt: 142 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005832
CAS Number: 14268-66-7
EINECS: 238-161-6
UN: 2811
ADR: 6.1,III