4-Chloro-2-nitrobenzoic acid
Supplier: Apollo Scientific
Warning
OR28593-100G OR28593-25G
APOSOR28593-100GEA 29 EUR
APOSOR28593-100G APOSOR28593-25G
4-Chloro-2-nitrobenzoic acid
4-Chloro-2-nitrobenzoic acid
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Melting Pt: 142…144 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00007214
CAS Number: 6280-88-2