Methyl-4-amino-3-methoxybenzoate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-amino-3-methoxybenzoate
OR2860-25G OR2860-5G
APOSOR2860-25GEA 109 EUR
APOSOR2860-25G APOSOR2860-5G
Methyl-4-amino-3-methoxybenzoate 98%
Methyl 4-amino-3-methoxybenzoate
Formula: C₉H₁₁NO₃
MW: 181.1900 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00017203
CAS Number: 41608-64-4