Thioacetamide 98%
Supplier: Apollo Scientific
Danger
APOSOR28604-25GEA 109 EUR
APOSOR28604-25G APOSOR28604-500G APOSOR28604-100G
Thioacetamide 98%
Thioacetamide
Formula: CH₃CSNH₂
MW: 75,13 g/mol
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00008070
EINECS: 200-541-4