Pyrrolidine-1-carbonitrile 97%
Supplier: Apollo Scientific
Danger
Synonyms: 1-Cyanopyrrolidine
OR28605-5G OR28605-100MG OR28605-25G
APOSOR28605-5GEA 226 EUR
APOSOR28605-5G APOSOR28605-100MG APOSOR28605-25G
Pyrrolidine-1-carbonitrile 97%
Pyrrolidine-1-carbonitrile
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 75…77 °C/1.8mmHg
Density: 0.954 g/cm³ (25 °C)
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00003165
CAS Number: 1530-88-7
EINECS: 216-234-3
UN: 3276
ADR: 6.1,III