2-Chloro-5-iodotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-4-iodo-2-methylbenzene
OR2862-10G OR2862-1G OR2862-5G
APOSOR2862-10GEA 48 EUR
APOSOR2862-10G APOSOR2862-1G APOSOR2862-5G
2-Chloro-5-iodotoluene
2-Chloro-5-iodotoluene
Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 239 °C
Density: 1.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD03093903
CAS Number: 116632-41-8