α-Chloro-o-xylene 98+%
Supplier: Apollo Scientific
|
Danger
|
OR28627-1KG
OR28627-500G
OR28627-5KG
APOSOR28627-1KGEA
563
EUR
APOSOR28627-1KG
APOSOR28627-500G
APOSOR28627-5KG
α-Chloro-o-xylene 98+%
Intermediate for organic synthesis.
Formula:
C₈H₉Cl Boiling Pt: 197…199 °C Melting Pt: 2 °C Density: 1.063 g/cm³ (25 °C) Flash Pt: 73.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000899 CAS Number: 552-45-4 EINECS: 209-013-8 UN: 3265 ADR: 8,II |