Ethyl cyanoacetate 98%
Supplier: Apollo Scientific
Warning
OR28668-5G OR28668-25G OR28668-100G
APOSOR28668-5GEA 36.4 EUR
APOSOR28668-5G APOSOR28668-25G APOSOR28668-100G
Ethyl cyanoacetate 98%
Ethyl cyanoacetate
Formula: C₅H₇NO₂
MW: 113.12 g/mol
Boiling Pt: 208…210 °C
Melting Pt: –23 °C
Density: 1.063 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00001940
CAS Number: 105-56-6
EINECS: 203-309-0
UN: 3334
ADR: 9,III