Maltol
Supplier: Apollo Scientific
Warning
Synonyms: 3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
OR28677-100G OR28677-25G OR28677-500G
APOSOR28677-100GEA 27.7 EUR
APOSOR28677-100G APOSOR28677-25G APOSOR28677-500G
Maltol
Maltol
Formula: C₆H₆O₃
MW: 126.11 g/mol
Boiling Pt: 170 °C/100mmHg
Melting Pt: 160…165 °C
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00006578
CAS Number: 118-71-8
EINECS: 204-271-8