2'-Aminoacetophenone 98%
Supplier: Apollo Scientific
Warning
Synonyms: 1-(2-Aminophenyl)ethanone, o-Aminoacetophenone
OR28679-100G OR28679-1KG OR28679-500G
APOSOR28679-100GEA 63.8 EUR
APOSOR28679-100G APOSOR28679-1KG APOSOR28679-500G
2'-Aminoacetophenone 98%
2'-Aminoacetophenone
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 253…255 °C
Melting Pt: 20
Density: 1.118 g/cm³ (25 °C)
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007717
CAS Number: 551-93-9
EINECS: 209-002-8
Merck Index: 13,00412