2-Chloro-6-iodotoluene
Supplier: Apollo Scientific
Warning
Synonyms: 1-Chloro-3-iodo-2-methylbenzene
OR2869-100G OR2869-1KG OR2869-500G
APOSOR2869-100GEA 79.9 EUR
APOSOR2869-100G APOSOR2869-1KG APOSOR2869-500G
2-Chloro-6-iodotoluene
2-Chloro-6-iodotoluene
Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 117…119 °C
Melting Pt: 11 °C
Density: 1.82 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00019013
CAS Number: 42048-11-3