1,3-Cyclohexanedione 99%
Supplier: Apollo Scientific
Warning
Synonyms: Cyclohexane-1,3-dione
OR28703-100G OR28703-1KG OR28703-500G
APOSOR28703-100GEA 21.9 EUR
APOSOR28703-100G APOSOR28703-1KG APOSOR28703-500G
1,3-Cyclohexanedione 99%
1,3-Cyclohexanedione
Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 235
Melting Pt: 101…105 °C
Density: 1.1 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00001585
CAS Number: 504-02-9
EINECS: 207-980-0
Merck Index: 13,03200