Ethyl-2-methylcyclopropanecarboxylate
Supplier: Apollo Scientific
Warning
OR28709-250MG OR28709-500MG
APOSOR28709-250MGEA 65.4 EUR
APOSOR28709-250MG APOSOR28709-500MG
Ethyl-2-methylcyclopropanecarboxylate
Ethyl-2-methylcyclopropanecarboxylate
Formula: C₇H₁₂O₂
MW: 128.17 g/mol
Boiling Pt: 77 °C/68.5mmHg
Density: 0.938 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Ambient
MDL Number: MFCD00009943
CAS Number: 20913-25-1
EINECS: 244-113-5
UN: 3272
ADR: 3,III