2-Mercapto-1-methylimidazole
Supplier: Apollo Scientific
Warning
Synonyms: 2-Mercapto-N-methyl-1H-imidazole, Thiamazole, Methimazole, 1-Methyl-1H-imidazole-2(3H)-thione, 2-Mercapto-N-methylimidazole, 2-Mercapto-1-methyl-1H-imidazole
OR28714-100G OR28714-25G OR28714-500G
APOSOR28714-100GEA 79.9 EUR
APOSOR28714-100G APOSOR28714-25G APOSOR28714-500G
2-Mercapto-1-methylimidazole
2-Mercapto-1-methylimidazole
Formula: C₄H₆N₂S
Boiling Pt: 280 °C
Melting Pt: 144…147 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00179321
CAS Number: 60-56-0
EINECS: 200-482-4