(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 98%
Supplier: Apollo Scientific
Danger
Synonyms: Naproxen, (S)-2-(6-Methoxynaphthalen-2-yl)propanoic acid, (S)-(+)-6-Methoxy-alpha-methyl-2-naphthaleneacetic Acid
OR28717-25G OR28717-100G OR28717-5G
APOSOR28717-25GEA 66.7 EUR
APOSOR28717-25G APOSOR28717-100G APOSOR28717-5G
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid 98%
(S)-(+)-2-(6-Methoxy-2-naphthyl)propionic acid
Formula: C₁₄H₁₄O₃
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00010500
CAS Number: 22204-53-1
UN: 2811
ADR: 6.1,III