Isochroman
Supplier: Apollo Scientific
Warning
Synonyms: 3,4-Dihydro-1H-2-benzopyran
OR28756-250G OR28756-50G
APOSOR28756-250GEA 109 EUR
APOSOR28756-250G APOSOR28756-50G
Isochroman
Isochroman
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 89…90 °C/12mmHg
Density: 1.067 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00006913
CAS Number: 493-05-0
EINECS: 207-774-0