m-Anisidine
Supplier: Apollo Scientific
Warning
OR28775-25G OR28775-5G
APOSOR28775-25GEA 29 EUR
APOSOR28775-25G APOSOR28775-5G
m-Anisidine
m-Anisidine
Formula: C₇H₉NO
Boiling Pt: 250…251 °C
Melting Pt: –1 °C
Density: 1.096 g/cm³ (25 °C)
Flash Pt: 126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007783
CAS Number: 536-90-3
EINECS: 208-651-4
UN: 2431
ADR: 6.1,III