2(5H)-Furanone 97%
Supplier: Apollo Scientific
Warning
OR2878-100G OR2878-25G OR2878-5G
APOSOR2878-100GEA 208 EUR
APOSOR2878-100G APOSOR2878-25G APOSOR2878-5G
2(5H)-Furanone 97%
2(5H)-Furanone
Formula: C₄H₄O₂
Boiling Pt: 212…214 °C/760mmHg
Melting Pt: 44320 °C
Density: 1.185 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005376
CAS Number: 497-23-4