3,5-Dimethyl-1H-pyrazole 99%
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Dimethylpyrazol, 3,5-Dimethyl-1H-pyrazole, 3,5-Dimethylpyrazole
OR28782-500G OR28782-1KG OR28782-100G
APOSOR28782-500GEA 36.4 EUR
APOSOR28782-500G APOSOR28782-1KG APOSOR28782-100G
3,5-Dimethyl-1H-pyrazole 99%
3,5-Dimethyl-1H-pyrazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 218 °C
Melting Pt: 105…108 °C
Storage Temperature: Ambient
MDL Number: MFCD00005243
CAS Number: 67-51-6
EINECS: 200-657-5