Clioquinol
Supplier: Apollo Scientific
Danger
Synonyms: 5-Chloro-7-iodo-8-quinolinol, 5-Chloro-8-hydroxy-7-iodoquinoline, Clioquinol
OR28788-1G OR28788-5G
APOSOR28788-1GEA 21.9 EUR
APOSOR28788-1G APOSOR28788-5G
Clioquinol
Clioquinol
Formula: C₉H₅ClINO
Melting Pt: 175 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00006787
CAS Number: 130-26-7
EINECS: 204-984-4
UN: 2811
ADR: 6.1,III