1-Amino-2-methylbutane
Supplier: Apollo Scientific
Danger
Synonyms: 2-Methylbutylamine, 2-Methyl-1-butylamine
OR28802-5G OR28802-1G OR28802-10G
APOSOR28802-5GEA 87 EUR
APOSOR28802-5G APOSOR28802-1G APOSOR28802-10G
1-Amino-2-methylbutane
1-Amino-2-methylbutane
Formula: C₅H₁₃N
Boiling Pt: 94…97 °C
Density: 0.739 g/cm³ (25 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00008147
CAS Number: 96-15-1
UN: 2733
ADR: 3+8,II