4-Acetyl-6-tert-butyl-1,1-dimethylindan
Supplier: Apollo Scientific
Warning
Synonyms: 4-Acetyl-6-(tert-butyl)-1,1,-dimethylindane
OR28804-1G OR28804-5G OR28804-25G
APOSOR28804-1GEA 21.9 EUR
APOSOR28804-1G APOSOR28804-5G APOSOR28804-25G
4-Acetyl-6-tert-butyl-1,1-dimethylindan
4-Acetyl-6-tert-butyl-1,1-dimethylindan
Formula: C₁₇H₂₄O
MW: 244.38 g/mol
Melting Pt: 76…79 °C
Storage Temperature: Ambient
MDL Number: MFCD00046324
CAS Number: 13171-00-1
EINECS: 236-114-4