4-Methyl-2-nitrophenol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Nitro-p-cresol, 1-Hydroxy-4-methyl-2-nitrobenzene
OR28809-1G OR28809-5G
APOSOR28809-1GEA 21.9 EUR
APOSOR28809-1G APOSOR28809-5G
4-Methyl-2-nitrophenol 98%
4-Methyl-2-nitrophenol
Formula: C₇H₇NO₃
Boiling Pt: 125 °C/22mmHg
Melting Pt: 32…35 °C
Density: 1.24 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00007120
CAS Number: 119-33-5
EINECS: 204-315-6
UN: 2446
ADR: 6.1,III