3,4-Diaminobenzoic acid 98%
Supplier: Apollo Scientific
Warning
OR28812-500G
APOSOR28812-500GEA 230 EUR
APOSOR28812-500G
3,4-Diaminobenzoic acid 98%
3,4-Diaminobenzoic acid
Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Melting Pt: 208…210 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007726
CAS Number: 619-05-6
EINECS: 210-577-2