Toluene-2-sulphonamide 98%
Supplier: Apollo Scientific
Warning
Synonyms: 2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide
OR28825-100G
APOSOR28825-100GEA 20.5 EUR
APOSOR28825-100G
Toluene-2-sulphonamide 98%
Toluene-2-sulphonamide
Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 143…145 °C/10mmHg
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00007934
CAS Number: 88-19-7
EINECS: 201-808-8