2,3-Dihydrobenzo[b]furan
Supplier: Apollo Scientific
|
Warning
|
OR28826-100G
OR28826-25G
OR28826-5G
APOSOR28826-100GEA
55.1
EUR
APOSOR28826-100G
APOSOR28826-25G
APOSOR28826-5G
2,3-Dihydrobenzo[b]furan
2,3-Dihydrobenzo[b]furan
Formula:
C₈H₈O Boiling Pt: 188…189 °C Melting Pt: –21 °C Density: 1.065 g/cm³ (25 °C) Flash Pt: 66 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005855 CAS Number: 496-16-2 EINECS: 207-817-3 |