4-(Dimethylamino)pyridine 99%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Dimethylaminopyridine, DMAP, N,N-Dimethylpyridin-4-amine
OR28827-100G OR28827-50G OR28827-250G
APOSOR28827-100GEA 27.7 EUR
APOSOR28827-100G APOSOR28827-50G APOSOR28827-250G
4-(Dimethylamino)pyridine 99%
4-(Dimethylamino)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 162 °C/50mmHg
Melting Pt: 110…113 °C
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00006418
CAS Number: 1122-58-3
EINECS: 214-353-5
UN: 2928
ADR: 6.1+8 I