2-Amino-4-nitrophenol
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-Hydroxy-5-nitroaniline
OR28831-100G
OR28831-25G
APOSOR28831-100GEA
63.8
EUR
APOSOR28831-100G
APOSOR28831-25G
2-Amino-4-nitrophenol
2-Amino-4-nitrophenol
Formula:
C₆H₆N₂O₃ Melting Pt: 140…145 °C Flash Pt: 100 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007695 CAS Number: 99-57-0 EINECS: 202-767-9 |