Adamantane
Supplier: Apollo Scientific
Warning
OR28833-100G OR28833-250G
APOSOR28833-100GEA 23.2 EUR
APOSOR28833-100G APOSOR28833-250G
Adamantane
Adamantane
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Melting Pt: 205…212 °C
Density: 1.07 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074719
CAS Number: 281-23-2
EINECS: 206-001-4
UN: 3077
ADR: 9+EHS,III