1-Adamantanol
Supplier: Apollo Scientific
Warning
Synonyms: 1-Tricyclo[3.3.1.1(3,7)]decanol, 1-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-1-ol, Adamantan-1-ol
OR28841-25G OR28841-100G OR28841-500G
APOSOR28841-25GEA 21.9 EUR
APOSOR28841-25G APOSOR28841-100G APOSOR28841-500G
1-Adamantanol
1-Adamantanol
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Melting Pt: 245…247 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00074729
CAS Number: 768-95-6
EINECS: 212-202-8