4-Amino-4H-1,2,4-triazole 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-1,2,4-triazole, 1,2,4-Triazol-4-ylamine, 4H-1,2,4-Triazol-4-amine
OR28852-100G OR28852-25G OR28852-500G
APOSOR28852-100GEA 24.8 EUR
APOSOR28852-100G APOSOR28852-25G APOSOR28852-500G
4-Amino-4H-1,2,4-triazole 98%
4-Amino-4H-1,2,4-triazole
Formula: C₂H₄N₄
MW: 84.08 g/mol
Melting Pt: 83…88 °C
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00003099
CAS Number: 584-13-4
EINECS: 209-533-5