4-(1H-Pyrazol-5-yl)aniline
Supplier: Apollo Scientific
Warning
OR2888-100MG OR2888-1G OR2888-250MG
APOSOR2888-100MGEA 36.4 EUR
APOSOR2888-100MG APOSOR2888-1G APOSOR2888-250MG
4-(1H-Pyrazol-5-yl)aniline
4-(1H-Pyrazol-5-yl)aniline
Formula: C₉H₉N₃
Melting Pt: 110…115 °C
Storage Temperature: Ambient
MDL Number: MFCD09055175
CAS Number: 89260-45-7